Product Details
- SNP ID
-
rs6060
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:154565984 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCCTGATGCTGGAGGCTGTCTTCA[C/T]GCTGACCCAGACCTGGTGGGTGCAC
- Phenotype
-
MIM: 134830
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FGB
PubMed Links
Gene Details
- Gene
- FGB
- Gene Name
- fibrinogen beta chain
View Full Product Details