Product Details

SNP ID
rs9510734
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:23426720 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGAAGCAAAAGCAAGAAGACTGGAA[A/G]TGAGTCAGACAGGTGATACACAAGT
Phenotype
MIM: 604490
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SACS PubMed Links

Gene Details

Gene
SACS
Gene Name
sacsin molecular chaperone
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278055.1 Intron NP_001264984.1
NM_014363.5 Intron NP_055178.3
XM_005266338.2 Intron XP_005266395.1
XM_011535039.2 Intron XP_011533341.1
XM_017020539.1 Intron XP_016876028.1
Gene
SACS-AS1
Gene Name
SACS antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details