Product Details

SNP ID
rs8177468
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.19:12738377 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GTAGGGACTGAGGCTCAATCCCACC[A/G]TAACCCATACTCTCCTCTCAAGGCC
Phenotype
MIM: 601913
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ASNA1 PubMed Links

Gene Details

Gene
ASNA1
Gene Name
arsA arsenite transporter, ATP-binding, homolog 1 (bacterial)
There are no transcripts associated with this gene.

Gene
C19orf43
Gene Name
chromosome 19 open reading frame 43
There are no transcripts associated with this gene.

View Full Product Details