Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001077238.1 | 410 | Intron | NP_001070706.1 | ||
NM_152988.2 | 410 | Intron | NP_694533.1 | ||
XM_011528138.2 | 410 | Intron | XP_011526440.1 | ||
XM_011528139.1 | 410 | Intron | XP_011526441.1 | ||
XM_011528140.1 | 410 | Intron | XP_011526442.1 | ||
XM_017027016.1 | 410 | Intron | XP_016882505.1 | ||
XM_017027017.1 | 410 | UTR 5 | XP_016882506.1 | ||
XM_017027018.1 | 410 | Intron | XP_016882507.1 | ||
XM_017027019.1 | 410 | UTR 5 | XP_016882508.1 |