Product Details

SNP ID
rs12611254
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:58454293 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCGTGGCGAGGAGCCCTGGGTTCCC[A/G]GTGGAAAGGACATGACCCTGGCCAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF324B PubMed Links

Gene Details

Gene
ZNF324B
Gene Name
zinc finger protein 324B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207395.2 318 Missense Mutation AGT,GGT S63G NP_997278.2
XM_005258915.4 318 Missense Mutation AGT,GGT S63G XP_005258972.1
XM_005258918.4 318 Missense Mutation AGT,GGT S63G XP_005258975.1

View Full Product Details