Product Details

SNP ID
rs10194593
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:191950586 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTATTATTTTGCTTTTCTGTGCA[C/G]TTGTCTTTAAATTTTTGTTGGGAAT
Phenotype
MIM: 605734
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TMEFF2 PubMed Links

Gene Details

Gene
TMEFF2
Gene Name
transmembrane protein with EGF like and two follistatin like domains 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001305134.1 Intron NP_001292063.1
NM_001305145.1 Intron NP_001292074.1
NM_016192.3 Intron NP_057276.2
XM_011510890.2 Intron XP_011509192.1
XM_017003739.1 Intron XP_016859228.1
XM_017003740.1 Intron XP_016859229.1

View Full Product Details