Product Details

SNP ID
rs3731897
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.2:219281929 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ATGCCCTAAGCTCTCTCCTCATCCT[A/G]CCTTTCCAGGAACTGGCCCATCTCG
Phenotype
MIM: 604139 MIM: 601773
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNAJB2 PubMed Links

Gene Details

Gene
DNAJB2
Gene Name
DnaJ heat shock protein family (Hsp40) member B2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039550.1 Intron NP_001034639.1
NM_006736.5 Intron NP_006727.2
Gene
PTPRN
Gene Name
protein tyrosine phosphatase, receptor type N
There are no transcripts associated with this gene.

Gene
TUBA4B
Gene Name
tubulin alpha 4b
There are no transcripts associated with this gene.

View Full Product Details