Product Details

SNP ID
rs6710951
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:219331283 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGGAGGCCTGGAATCATTAGAAAA[A/G]GTATTGGTTTGTTTAGGGTTTGAGG
Phenotype
MIM: 612721
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RESP18 PubMed Links

Gene Details

Gene
RESP18
Gene Name
regulated endocrine specific protein 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007089.3 Intron NP_001007090.3
XM_011511165.2 Intron XP_011509467.1
XM_011511167.2 Intron XP_011509469.1
XM_017004079.1 Intron XP_016859568.1

View Full Product Details