Product Details

SNP ID
rs7248632
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.19:13884713 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CAGAACAGGTGCCACTCTCAGACAC[A/G]GGCAGTACAGGTGGCCAGGCGGGTA
Phenotype
MIM: 612746 MIM: 608229
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C19orf57 PubMed Links

Gene Details

Gene
C19orf57
Gene Name
chromosome 19 open reading frame 57
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024323.3 2766 Intron NP_077299.3
XM_006722885.3 2766 Intron XP_006722948.2
XM_011528286.2 2766 UTR 3 XP_011526588.1
XM_011528287.2 2766 UTR 3 XP_011526589.1
XM_017027300.1 2766 Intron XP_016882789.1
XM_017027301.1 2766 Intron XP_016882790.1
Gene
MIR181C
Gene Name
microRNA 181c
There are no transcripts associated with this gene.

Gene
MIR181D
Gene Name
microRNA 181d
There are no transcripts associated with this gene.

Gene
NANOS3
Gene Name
nanos C2HC-type zinc finger 3
There are no transcripts associated with this gene.

View Full Product Details