Product Details

SNP ID
rs11879596
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:11426557 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCATGGAGTCCAGCCGGTTCTCCAA[A/G]TTGAGGCTCTCGTCCTGGGGCGTGG
Phenotype
MIM: 615956 MIM: 177060 MIM: 616743
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC151 PubMed Links

Gene Details

Gene
CCDC151
Gene Name
coiled-coil domain containing 151
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001302453.1 870 Silent Mutation AAC,AAT N189N NP_001289382.1
NM_001302454.1 870 Silent Mutation AAC,AAT N183N NP_001289383.1
NM_145045.4 870 Silent Mutation AAC,AAT N243N NP_659482.3
XM_017026241.1 870 Silent Mutation AAC,AAT N243N XP_016881730.1
Gene
PRKCSH
Gene Name
protein kinase C substrate 80K-H
There are no transcripts associated with this gene.

Gene
RGL3
Gene Name
ral guanine nucleotide dissociation stimulator like 3
There are no transcripts associated with this gene.

View Full Product Details