Product Details

SNP ID
rs12982049
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:19669400 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACGACCCCGTCCCCCAGCTTCGCT[C/G]CTCCCAGGGCTGCAGTAAATCCCTA
Phenotype
MIM: 603983
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ATP13A1 PubMed Links

Gene Details

Gene
ATP13A1
Gene Name
ATPase 13A1
There are no transcripts associated with this gene.

Gene
ZNF101
Gene Name
zinc finger protein 101
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300949.1 Intron NP_001287878.1
NM_033204.3 Intron NP_149981.2

View Full Product Details