Product Details

SNP ID
rs6720523
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:218672452 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGTCTGGGAGTGGCAGGAACTTG[A/G]GGGAGACTCAAAGGAGCAGGGTTGG
Phenotype
MIM: 603647 MIM: 616014 MIM: 607652
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BCS1L PubMed Links

Gene Details

Gene
BCS1L
Gene Name
BCS1 homolog, ubiquinol-cytochrome c reductase complex chaperone
There are no transcripts associated with this gene.

Gene
RNF25
Gene Name
ring finger protein 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022453.2 Intron NP_071898.2
XM_017004695.1 Intron XP_016860184.1
Gene
STK36
Gene Name
serine/threonine kinase 36
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243313.1 Intron NP_001230242.1
NM_015690.4 Intron NP_056505.2
XM_005246464.1 Intron XP_005246521.1
XM_011510959.1 Intron XP_011509261.1
XM_011510960.1 Intron XP_011509262.1
XM_011510961.1 Intron XP_011509263.1
XM_017003804.1 Intron XP_016859293.1
XM_017003805.1 Intron XP_016859294.1

View Full Product Details