Product Details

SNP ID
rs11685653
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:240446167 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGTCTGCGCCCAGGAGGGCCCAC[G/T]GTGGCCTAGGGTCCGAAACGGGGGA
Phenotype
MIM: 600395 MIM: 615209
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GPC1 PubMed Links

Gene Details

Gene
GPC1
Gene Name
glypican 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002081.2 Intron NP_002072.2
XM_011510976.2 Intron XP_011509278.1
Gene
MIR149
Gene Name
microRNA 149
There are no transcripts associated with this gene.

Gene
PP14571
Gene Name
uncharacterized LOC100130449
There are no transcripts associated with this gene.

View Full Product Details