Product Details

SNP ID
rs10933603
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:240444317 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAAGTACAACGCTGCTAGGCTTCTG[C/T]TCTCGCCCCACAGAAGTTCAGGTTC
Phenotype
MIM: 600395
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPC1 PubMed Links

Gene Details

Gene
GPC1
Gene Name
glypican 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002081.2 Intron NP_002072.2
XM_011510976.2 Intron XP_011509278.1
Gene
PP14571
Gene Name
uncharacterized LOC100130449
There are no transcripts associated with this gene.

View Full Product Details