Product Details

SNP ID
rs11795584
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:71668433 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTGAATACTGACGACTCTGATTC[C/T]GCAGATGAGAGCAGCGACTTACCGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC100129291 PubMed Links

Gene Details

Gene
LOC100129291
Gene Name
chromosome X open reading frame 49 pseudogene
There are no transcripts associated with this gene.

Gene
LOC101059915
Gene Name
chromosome X open reading frame 49-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001323075.1 Intron NP_001310004.1

View Full Product Details