Product Details
- SNP ID
-
rs35416933
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:241688523 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- TGCATACCTAGAGTAGCTTTATGTT[C/T]TCCAGACTGAAGCCTCCCCAATTCA
- Phenotype
-
MIM: 188345
MIM: 608525
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
DTYMK
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs146565733] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DTYMK
- Gene Name
- deoxythymidylate kinase
There are no transcripts associated with this gene.
- Gene
- ING5
- Gene Name
- inhibitor of growth family member 5
View Full Product Details