Product Details

SNP ID
rs35471768
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:68005101 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTCACAAGCAGTAAATACATCCA[C/T]AGTCACCTCTGTTTTGTCTTTCTGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM19A1 PubMed Links

Gene Details

Gene
FAM19A1
Gene Name
family with sequence similarity 19 member A1, C-C motif chemokine like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001252216.1 Intron NP_001239145.1
NM_213609.3 Intron NP_998774.2
XM_017006386.1 Intron XP_016861875.1
XM_017006387.1 Intron XP_016861876.1
XM_017006388.1 Intron XP_016861877.1
XM_017006389.1 Intron XP_016861878.1
XM_017006390.1 Intron XP_016861879.1

View Full Product Details