Product Details

SNP ID
rs2070581
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.2:73893054 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GAGAAGAACCTCTCATACCCTCGGT[A/G]AGTACTGTACGGCTTTTGCCACCTC
Phenotype
MIM: 102545
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACTG2 PubMed Links

Gene Details

Gene
ACTG2
Gene Name
actin, gamma 2, smooth muscle, enteric
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199893.1 Intron NP_001186822.1
NM_001615.3 Intron NP_001606.1

View Full Product Details