Product Details

SNP ID
rs1135251
Assay Type
Validated
NCBI dbSNP Submissions
57
Location
Chr.1:26051511 on Build GRCh38
Set Membership
HapMap JSNP Validated
Context Sequence [VIC/FAM]
TTTCTGACAATCGCAGGTCACCCAA[C/T]GACCAGGCATTATTATACAAAAGTT
Phenotype
MIM: 609617 MIM: 606131
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC30A2 PubMed Links

Gene Details

Gene
SLC30A2
Gene Name
solute carrier family 30 member 2
There are no transcripts associated with this gene.

Gene
TRIM63
Gene Name
tripartite motif containing 63
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032588.3 1564 UTR 3 NP_115977.2
XM_017002559.1 1564 Intron XP_016858048.1

View Full Product Details