Product Details

SNP ID
rs6665733
Assay Type
Functionally tested
NCBI dbSNP Submissions
41
Location
Chr.1:23873668 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTGCTTATTATATGTAATAGGGG[C/T]GTGGAGGAGGCAAGGGAGCTGAATT
Phenotype
MIM: 605051 MIM: 612280
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CNR2 PubMed Links

Gene Details

Gene
CNR2
Gene Name
cannabinoid receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001841.2 2360 Intron NP_001832.1
XM_011540629.2 2360 UTR 3 XP_011538931.1
XM_017000261.1 2360 UTR 3 XP_016855750.1
Gene
FUCA1
Gene Name
fucosidase, alpha-L- 1, tissue
There are no transcripts associated with this gene.

View Full Product Details