Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001040451.2 | Intron | NP_001035541.1 | |||
NM_001040452.2 | Intron | NP_001035542.1 | |||
NM_025158.4 | Intron | NP_079434.3 | |||
XM_005265993.4 | Intron | XP_005266050.1 | |||
XM_006714921.3 | Intron | XP_006714984.2 | |||
XM_006714922.3 | Intron | XP_006714985.1 | |||
XM_017009891.1 | Intron | XP_016865380.1 | |||
XM_017009892.1 | Intron | XP_016865381.1 | |||
XM_017009893.1 | Intron | XP_016865382.1 | |||
XM_017009894.1 | Intron | XP_016865383.1 | |||
XM_017009895.1 | Intron | XP_016865384.1 |