Product Details

SNP ID
rs2961663
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:178186543 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGAGCTGTTTCATGACATTTATAC[C/T]GAGTTCTTTAAAAAGTTTAACATTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GMCL1P1 PubMed Links

Gene Details

Gene
GMCL1P1
Gene Name
germ cell-less, spermatogenesis associated 1 pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details