Product Details

SNP ID
rs4401691
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.6:157299382 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
GTATCAGAGTTCAGATTCTTCAAAG[C/G]GACACCAGCTATGTTCACTCCACTC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LDHAL6FP PubMed Links

Gene Details

Gene
LDHAL6FP
Gene Name
lactate dehydrogenase A like 6F, pseudogene
There are no transcripts associated with this gene.

Gene
TMEM242
Gene Name
transmembrane protein 242
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018452.5 Intron NP_060922.2

View Full Product Details