Product Details

SNP ID
rs6455606
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:158852488 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACACTCCTAACCCTACTACAACACC[C/T]GCTCTCAAACAGAGAAGGGAAAGGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OSTCP1 PubMed Links

Gene Details

Gene
OSTCP1
Gene Name
oligosaccharyltransferase complex subunit pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details