Product Details

SNP ID
rs11734833
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:100186912 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGGAAGAGTAATCAACTACACACA[A/G]AAATATCAGTTCAATATTTTGCATT
Phenotype
MIM: 607730
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DDIT4L PubMed Links

Gene Details

Gene
DDIT4L
Gene Name
DNA damage inducible transcript 4 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145244.3 1592 UTR 3 NP_660287.1
Gene
LOC101929353
Gene Name
uncharacterized LOC101929353
There are no transcripts associated with this gene.

View Full Product Details