Product Details
- SNP ID
-
rs11706973
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:196579098 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATTTTTTATTCTTTTCATCTTAGGC[G/T]TAAGGCATACTCATTTGACTCAAGT
- Phenotype
-
MIM: 609112
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
FBXO45
PubMed Links
Gene Details
- Gene
- FBXO45
- Gene Name
- F-box protein 45
View Full Product Details