Product Details

SNP ID
rs10001403
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:83298925 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTAAGCGTACAGACTGTGCACTCA[A/T]AAACTTATCTAGGATACTATTTTGC
Phenotype
MIM: 604724
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
HPSE PubMed Links

Gene Details

Gene
HPSE
Gene Name
heparanase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098540.2 Intron NP_001092010.1
NM_001166498.2 Intron NP_001159970.1
NM_001199830.1 Intron NP_001186759.1
NM_006665.5 Intron NP_006656.2

View Full Product Details