Product Details

SNP ID
rs35191642
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:108811753 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTTCAAAATGCTAAACTGGCTTA[A/G]GGGTGTGGAATGTTACCAGAAATGT
Phenotype
MIM: 610004
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COL25A1 PubMed Links

Gene Details

Gene
COL25A1
Gene Name
collagen type XXV alpha 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256074.1 4584 Intron NP_001243003.1
NM_032518.2 4584 Intron NP_115907.2
NM_198721.3 4584 UTR 3 NP_942014.1
XM_011532333.2 4584 UTR 3 XP_011530635.1
XM_011532334.2 4584 UTR 3 XP_011530636.1
XM_011532335.2 4584 UTR 3 XP_011530637.1
XM_011532338.2 4584 UTR 3 XP_011530640.1
XM_011532355.2 4584 UTR 3 XP_011530657.1
XM_011532356.2 4584 UTR 3 XP_011530658.1
XM_011532358.2 4584 UTR 3 XP_011530660.1
XM_017008735.1 4584 UTR 3 XP_016864224.1
XM_017008736.1 4584 UTR 3 XP_016864225.1
XM_017008737.1 4584 UTR 3 XP_016864226.1

View Full Product Details