Product Details

SNP ID
rs35346847
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.4:139466401 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
GTGAATATAATTTGCTTAAAACATT[A/T]TAAGTCATTTTTCATTGATGAAGTG
Phenotype
MIM: 605950
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
RAB33B PubMed Links

Gene Details

Gene
RAB33B
Gene Name
RAB33B, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031296.2 Intron NP_112586.1
XM_011532299.1 Intron XP_011530601.1

View Full Product Details