Product Details

SNP ID
rs1958260
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:35534869 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGCGCCGCGGCAAGGCACCCACGG[A/G]CTGCGCGTCTGGACCCGCGGCCGCG
Phenotype
MIM: 614027 MIM: 608884
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
INSM2 PubMed Links

Gene Details

Gene
INSM2
Gene Name
INSM transcriptional repressor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032594.3 828 Missense Mutation GAC,GGC D206G NP_115983.3
Gene
RALGAPA1
Gene Name
Ral GTPase activating protein catalytic alpha subunit 1
There are no transcripts associated with this gene.

View Full Product Details