Product Details

SNP ID
rs9396
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:90515792 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATACACACAATCACAGAATTAAA[A/T]ATCTTTACTCCTTAAAAAAAAATGA
Phenotype
MIM: 602614
Polymorphism
A/T, Transversion Substitution
Allele Nomenclature
Literature Links
MAP3K7 PubMed Links

Gene Details

Gene
MAP3K7
Gene Name
mitogen-activated protein kinase kinase kinase 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003188.3 2736 UTR 3 NP_003179.1
NM_145331.2 2736 UTR 3 NP_663304.1
NM_145332.2 2736 UTR 3 NP_663305.1
NM_145333.2 2736 UTR 3 NP_663306.1
XM_006715553.2 2736 UTR 3 XP_006715616.1
XM_017011226.1 2736 UTR 3 XP_016866715.1

View Full Product Details