Product Details
- SNP ID
-
rs9901910
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:82240131 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGTCCCCATCTGCCCCTGGCCTTGC[C/G]CCAGCTGGGCTTGAGCTCTTGCAGC
- Phenotype
-
MIM: 600864
MIM: 603877
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CSNK1D
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs58378835] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CSNK1D
- Gene Name
- casein kinase 1 delta
- Gene
- MIR6787
- Gene Name
- microRNA 6787
There are no transcripts associated with this gene.
- Gene
- SLC16A3
- Gene Name
- solute carrier family 16 member 3
View Full Product Details