Product Details

SNP ID
rs2569
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:74406153 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACATTGGTGCTGCCCCCCGCCTA[A/C]CTGGAGATGTCTCTAAAATTCTGGA
Phenotype
MIM: 609999
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SYNDIG1L PubMed Links

Gene Details

Gene
SYNDIG1L
Gene Name
synapse differentiation inducing 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105579.1 4933 UTR 3 NP_001099049.1
XM_017021600.1 4933 UTR 3 XP_016877089.1

View Full Product Details