Product Details

SNP ID
rs2051618
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:87520868 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGACAGAGTATTTAATTATGAAAT[A/C]ATATATATAGGTATTTGCAATTGCT
Phenotype
MIM: 300348
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
KLHL4 PubMed Links

Gene Details

Gene
KLHL4
Gene Name
kelch like family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019117.4 Intron NP_061990.2
NM_057162.2 Intron NP_476503.1

View Full Product Details