Product Details

SNP ID
rs2748967
Assay Type
Functionally tested
NCBI dbSNP Submissions
35
Location
Chr.1:1929904 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCACATTGGCCATGGATCCTTCCT[C/T]GGCCCTGCTCGGGTCTGAGGGTCAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CFAP74 PubMed Links

Gene Details

Gene
CFAP74
Gene Name
cilia and flagella associated protein 74
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304360.1 Intron NP_001291289.1
XM_011542333.2 Intron XP_011540635.2
XM_011542335.1 Intron XP_011540637.1
XM_011542336.2 Intron XP_011540638.1
XM_017002641.1 Intron XP_016858130.1
XM_017002642.1 Intron XP_016858131.1
XM_017002643.1 Intron XP_016858132.1
XM_017002644.1 Intron XP_016858133.1

View Full Product Details