Product Details

SNP ID
rs4648609
Assay Type
Validated
NCBI dbSNP Submissions
43
Location
Chr.1:1671097 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
ATCCTTCCTGCCATCGTGACACCAG[G/C]ACACAGATGGCCACAAGGAAGCAGA
Phenotype
Polymorphism
G/C, Transversion substitution
Allele Nomenclature
Literature Links
SLC35E2B PubMed Links

Gene Details

Gene
SLC35E2B
Gene Name
solute carrier family 35 member E2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001110781.2 Intron NP_001104251.1
NM_001290264.1 Intron NP_001277193.1
XM_011542072.1 Intron XP_011540374.1

View Full Product Details