Product Details

SNP ID
rs1886889
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:53191654 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAAAAATCAGGGAATTATGCAGTA[G/T]ATGGGTTAGACAGGAAAACATTCTA
Phenotype
MIM: 314690
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
KDM5C PubMed Links

Gene Details

Gene
KDM5C
Gene Name
lysine demethylase 5C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146702.1 6529 UTR 3 NP_001140174.1
NM_001282622.1 6529 UTR 3 NP_001269551.1
NM_004187.3 6529 UTR 3 NP_004178.2
XM_005262035.4 6529 Intron XP_005262092.1
XM_011530824.2 6529 Intron XP_011529126.1
XM_011530825.2 6529 Intron XP_011529127.1
XM_011530826.2 6529 Intron XP_011529128.1
XM_011530827.2 6529 Intron XP_011529129.1
XM_011530828.1 6529 Intron XP_011529130.1
XM_011530829.2 6529 Intron XP_011529131.1
XM_011530830.2 6529 Intron XP_011529132.1
XM_011530831.2 6529 Intron XP_011529133.1
XM_017029886.1 6529 Intron XP_016885375.1
XM_017029887.1 6529 Intron XP_016885376.1
XM_017029888.1 6529 Intron XP_016885377.1
XM_017029889.1 6529 Intron XP_016885378.1
Gene
MIR6894
Gene Name
microRNA 6894
There are no transcripts associated with this gene.

Gene
MIR6895
Gene Name
microRNA 6895
There are no transcripts associated with this gene.

View Full Product Details