Product Details

SNP ID
rs7553095
Assay Type
Functionally tested
NCBI dbSNP Submissions
38
Location
Chr.1:109558609 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCTCAGGGTCTGTGCTTCATCATC[A/T]CCCTGGCTTTTTTTTTGTGCTAATT
Phenotype
MIM: 139370
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
GNAI3 PubMed Links

Gene Details

Gene
GNAI3
Gene Name
G protein subunit alpha i3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006496.3 Intron NP_006487.1

View Full Product Details