Product Details
- SNP ID
-
rs1925781
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:83151519 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AAGACCATTAAGCCGCTTAACTCAT[A/C]GTGAGAAATTAGATCGCATTAGTTT
- Phenotype
-
MIM: 616823
MIM: 172100
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
DOPEY1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116798329] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DOPEY1
- Gene Name
- dopey family member 1
- Gene
- PGM3
- Gene Name
- phosphoglucomutase 3
View Full Product Details