Product Details

SNP ID
rs14661
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:103217107 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTGTACCAAAATCAGTGCCATTG[G/T]TGTGTGTGTACGTAACTATATACAT
Phenotype
MIM: 300692
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
BEX4 PubMed Links

Gene Details

Gene
BEX4
Gene Name
brain expressed X-linked 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080425.3 1106 UTR 3 NP_001073894.1
NM_001127688.2 1106 UTR 3 NP_001121160.1

View Full Product Details