Product Details

SNP ID
rs14518
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:79361170 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAGCATTTATCAATGGCACGTTT[T/G]TTGAAACCTGTAAGGAAATGTGCAA
Phenotype
MIM: 300222
Polymorphism
T/G, Transversion substitution
Allele Nomenclature
Literature Links
ITM2A PubMed Links

Gene Details

Gene
ITM2A
Gene Name
integral membrane protein 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171581.1 1048 Missense Mutation AAA,AAC K193N NP_001165052.1
NM_004867.4 1048 Missense Mutation AAA,AAC K237N NP_004858.1

View Full Product Details