Product Details

SNP ID
rs12946086
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:59220877 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGAAGAATGGGGTTGTGTGAATTGA[A/G]GGCTCTTCCGGAGGTGAAGGCTGTG
Phenotype
MIM: 616317 MIM: 613175
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GDPD1 PubMed Links

Gene Details

Gene
GDPD1
Gene Name
glycerophosphodiester phosphodiesterase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001165993.1 Intron NP_001159465.1
NM_001165994.1 Intron NP_001159466.1
NM_182569.3 Intron NP_872375.2
XM_017024521.1 Intron XP_016880010.1
Gene
SMG8
Gene Name
SMG8, nonsense mediated mRNA decay factor
There are no transcripts associated with this gene.

View Full Product Details