Product Details

SNP ID
rs823091
Assay Type
Functionally tested
NCBI dbSNP Submissions
63
Location
Chr.1:205823708 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTGCCTAGCTAACTCTTACTCAAC[C/T]TTGAAGTCTCAGCTTCAGTCATGAC
Phenotype
MIM: 610801
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PM20D1 PubMed Links

Gene Details

Gene
PM20D1
Gene Name
peptidase M20 domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152491.4 Intron NP_689704.4
XM_017000396.1 Intron XP_016855885.1
XM_017000397.1 Intron XP_016855886.1
XM_017000398.1 Intron XP_016855887.1
Gene
SLC41A1
Gene Name
solute carrier family 41 member 1
There are no transcripts associated with this gene.

View Full Product Details