Product Details
- SNP ID
-
rs12122693
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
50
- Location
-
Chr.1:204081798 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AACCTCTCCCCACCCTCCAGCCCTA[C/T]TGAAGGAGTCTGAAGTCACTCAACC
- Phenotype
-
MIM: 604748
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SOX13
PubMed Links
Gene Details
- Gene
- SOX13
- Gene Name
- SRY-box 13
View Full Product Details