Product Details

SNP ID
rs3922999
Assay Type
Validated
NCBI dbSNP Submissions
73
Location
Chr.1:202122532 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CCAGCTGAGTGCTAGCACCTAGTAA[A/G]TGTTCAGTAAGTGACAGTGATGGTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPR37L1 PubMed Links

Gene Details

Gene
GPR37L1
Gene Name
G protein-coupled receptor 37 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004767.3 Intron NP_004758.3
XM_011510158.2 Intron XP_011508460.1

View Full Product Details