Product Details

SNP ID
rs14762
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:20032735 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAACATGCAACCATCATCTTCCA[A/C]AGTCAAGTCACAATGTCAAATATTT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LAPTM4A PubMed Links

Gene Details

Gene
LAPTM4A
Gene Name
lysosomal protein transmembrane 4 alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014713.4 1680 UTR 3 NP_055528.1

View Full Product Details