Product Details

SNP ID
rs1868783
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.7:134290335 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TTTCCCCTTATTTGCTTTCTTGCCA[A/G]CCCGCTGATGAGGAGTGGGAACAGG
Phenotype
MIM: 610923
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC35B4 PubMed Links

Gene Details

Gene
SLC35B4
Gene Name
solute carrier family 35 member B4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032826.4 5818 UTR 3 NP_116215.1
XM_011516645.2 5818 Intron XP_011514947.1

View Full Product Details