Product Details

SNP ID
rs11653611
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:48726176 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAATCATCGTATTAAGGATGGGTG[C/T]TTCCAAGACCAGTGGGTACACCCTA
Phenotype
MIM: 604607 MIM: 609819 MIM: 610787
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HOXB13 PubMed Links

Gene Details

Gene
HOXB13
Gene Name
homeobox B13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006361.5 1625 UTR 3 NP_006352.2
Gene
MIR3185
Gene Name
microRNA 3185
There are no transcripts associated with this gene.

Gene
PRAC1
Gene Name
prostate cancer susceptibility candidate 1
There are no transcripts associated with this gene.

Gene
PRAC2
Gene Name
prostate cancer susceptibility candidate 2
There are no transcripts associated with this gene.

View Full Product Details