Product Details

SNP ID
rs2157840
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.17:45074105 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
CATTGCGCTCTATTTCTAGAAATAA[G/T]TACTGAAGTTAGTTTTCCATCTCCT
Phenotype
MIM: 160993
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
NMT1 PubMed Links

Gene Details

Gene
NMT1
Gene Name
N-myristoyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021079.4 Intron NP_066565.1
XM_011524858.2 Intron XP_011523160.1

View Full Product Details