Product Details
- SNP ID
-
rs2494519
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
47
- Location
-
Chr.1:159827283 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGTGAGGACCCGGCAGTGTTAGCA[C/T]GCAGAAACTCTGCAGAGTTTGAGGA
- Phenotype
-
MIM: 606620
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
C1orf204
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs182618093] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C1orf204
- Gene Name
- chromosome 1 open reading frame 204
There are no transcripts associated with this gene.
- Gene
- SLAMF8
- Gene Name
- SLAM family member 8
View Full Product Details